Assay Design



Gene Transcripts

Gene Symbol
Ern2
Entrez Gene ID
26918
Gene Name
endoplasmic reticulum (ER) to nucleus signalling 2
Gene Aliases
Ern1, Ire1, Ire1b
Chromosome Location
Chr.7: 122169745 - 122186226 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012016.2 NP_036146.2 5 - 6 438 62
GenBank mRNA AF071777.1 - 5 - 6 397 62
AK139291.1 - 5 - 6 430 62
AK153659.1 - 5 - 6 428 62
BC108999.2 - 5 - 6 397 62
BC109000.1 - 5 - 6 397 62

View Full Product Details