Assay Design



Gene Transcripts

Gene Symbol
Mki67
Entrez Gene ID
17345
Gene Name
antigen identified by monoclonal antibody Ki 67
Gene Aliases
D630048A14Rik, Ki-67, Ki67
Chromosome Location
Chr.7: 135689782 - 135716408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081117.2 NP_001074586.2 4 - 5 593 80
GenBank mRNA AK052772.1 - 4 - 5 513 80
AK082099.1 - 4 - 5 511 80
AK143677.1 - 4 - 5 632 80

View Full Product Details