Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146200.1 | NP_666312.1 | 5 - 6 | 539 | 92 |
GenBank mRNA | AK013160.1 | - | 5 - 6 | 566 | 92 |
AK076165.1 | - | 5 - 6 | 557 | 92 | |
AK088076.1 | - | 5 - 6 | 582 | 92 | |
AK152693.1 | - | 5 - 6 | 557 | 92 | |
AK160078.1 | - | 5 - 6 | 559 | 92 | |
AK166743.1 | - | 5 - 6 | 582 | 92 | |
AK168160.1 | - | 5 - 6 | 543 | 92 | |
AK168174.1 | - | 5 - 6 | 543 | 92 | |
BC024855.1 | - | 5 - 6 | 539 | 92 | |
BC025032.1 | - | 5 - 6 | 560 | 92 |