Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175103.3 | NP_780312.1 | 2 - 3 | 196 | 69 |
GenBank mRNA | AK002662.1 | - | 2 - 3 | 199 | 69 |
AK003922.1 | - | 2 - 3 | 199 | 69 | |
AK028165.1 | - | 2 - 3 | 210 | 69 | |
BC045523.1 | - | 2 - 3 | 214 | 69 |