Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013733.3 | NP_038761.1 | 7 - 8 | 1410 | 111 |
GenBank mRNA | AJ132771.2 | - | 7 - 8 | 1314 | 111 |
AK034839.1 | - | 7 - 8 | 1411 | 111 | |
AK132551.1 | - | - | 1234 | 111 | |
AK161016.1 | - | - | 238 | 111 | |
BC053740.1 | - | 7 - 8 | 1381 | 111 |