Assay Design



Gene Transcripts

Gene Symbol
Znrd1as
Entrez Gene ID
76416
Gene Name
Znrd1 antisense
Gene Aliases
1700022C21Rik, AI746295, Znrd1-as
Chromosome Location
Chr.17: 36958592 - 36965625 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029602.1 NP_083878.1 1 - 2 116 86
GenBank mRNA AK006236.1 - 1 - 2 109 86
BC027007.1 - 1 - 2 116 86

View Full Product Details