Assay Design



Gene Transcripts

Gene Symbol
Dnajc9
Entrez Gene ID
108671
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 9
Gene Aliases
5330419I01Rik, AU020082
Chromosome Location
Chr.14: 20384638 - 20388910 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134081.5 NP_598842.1 1 - 2 309 74
GenBank mRNA AK043725.1 - 1 - 2 279 74
AK075979.1 - 1 - 2 270 74
AK077324.1 - 1 - 2 279 74
AK153033.1 - 1 - 2 269 74
AK162075.1 - 1 - 2 184 74
BC014686.1 - 1 - 2 271 74
BC023787.1 - 1 - 2 267 74
BC027012.1 - 1 - 2 262 74
CX238858.1 - 1 - 2 309 74

View Full Product Details