Assay Design



Gene Transcripts

Gene Symbol
Kif6
Entrez Gene ID
319991
Gene Name
kinesin family member 6
Gene Aliases
D130004B10Rik, D130084M03Rik
Chromosome Location
Chr.17: 49615104 - 49909847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177052.3 NP_796026.2 3 - 4 317 70
GenBank mRNA AK043404.1 - 3 - 4 274 70
AK083760.1 - 3 - 4 354 70

View Full Product Details