Assay Design



Gene Transcripts

Gene Symbol
Trim31
Entrez Gene ID
224762
Gene Name
tripartite motif-containing 31
Gene Aliases
BC026666, HCG1, HCGI, RNF
Chromosome Location
Chr.17: 36898130 - 36910217 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146077.2 NP_666189.1 4 - 5 985 90
GenBank mRNA AK033650.1 - 4 - 5 998 90
BC026666.1 - 4 - 5 985 90

View Full Product Details