Assay Design
Gene Transcripts
- Gene Symbol
- Rrp36
- Entrez Gene ID
-
224823
- Gene Name
- ribosomal RNA processing 36 homolog (S. cerevisiae)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 46667442 - 46674451 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144857.1 |
NP_659106.1 |
2 - 3 |
204 |
65 |
GenBank mRNA |
BC011248.1 |
- |
2 - 3 |
204 |
65 |
View Full Product Details