Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010633.3 | NP_034763.3 | 3 - 4 | 906 | 108 |
GenBank mRNA | AK013347.1 | - | 3 - 4 | 911 | 108 |
AK030152.1 | - | - | 723 | 108 | |
AK218845.1 | - | 1 - 2 | 121 | 108 | |
BC058732.1 | - | 3 - 4 | 906 | 108 | |
X82320.1 | - | 2 - 3 | 392 | 108 | |
Y10725.2 | - | 3 - 4 | 763 | 108 |