Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153457.7 | NP_703187.2 | 1 - 2 | 744 | 133 |
GenBank mRNA | AB074899.1 | - | 1 - 2 | 241 | 133 |
AK132364.1 | - | 1 - 2 | 744 | 133 | |
BC030455.1 | - | 1 - 2 | 296 | 133 | |
BC053926.1 | - | 1 - 2 | 372 | 133 | |
BC058579.1 | - | 1 - 2 | 406 | 133 | |
CT010392.1 | - | 1 - 2 | 241 | 133 |