Assay Design



Gene Transcripts

Gene Symbol
Crhbp
Entrez Gene ID
12919
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
CRH-BP
Chromosome Location
Chr.13: 95431376 - 95444831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198408.3 NP_940800.1 1 - 2 234 79
GenBank mRNA AK135067.1 - 1 - 2 235 79
BC061247.1 - 1 - 2 175 79

View Full Product Details