Assay Design



Gene Transcripts

Gene Symbol
Pappa2
Entrez Gene ID
23850
Gene Name
pappalysin 2
Gene Aliases
PAPP-A2, PLAC3, Pappe
Chromosome Location
Chr.1: 158711727 - 158980523 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085376.2 NP_001078845.1 20 - 21 5191 60
GenBank mRNA BC094560.1 - 10 - 11 1451 60

View Full Product Details