Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013675.3 | NP_038703.3 | 35 - 36 | 7485 | 62 |
GenBank mRNA | AK147299.1 | - | 35 - 36 | 7486 | 62 |
AK147684.1 | - | 35 - 36 | 7078 | 62 | |
AK147704.1 | - | 35 - 36 | 7078 | 62 | |
AK220551.1 | - | 16 - 17 | 2585 | 62 | |
BC117711.2 | - | 18 - 19 | 2739 | 62 | |
BC129806.1 | - | 34 - 35 | 6818 | 62 | |
BC150783.1 | - | 34 - 35 | 6851 | 62 |