Assay Design



Gene Transcripts

Gene Symbol
Rc3h1
Entrez Gene ID
381305
Gene Name
RING CCCH (C3H) domains 1
Gene Aliases
5730557L09Rik, Gm551, N28103, mKIAA2025
Chromosome Location
Chr.1: 160906411 - 160974976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024952.2 NP_001020123.1 12 - 13 2642 60
GenBank mRNA AK139648.1 - 12 - 13 2614 60
AK173335.1 - 12 - 13 2588 60
AY948287.1 - 12 - 13 2583 60
BC138663.1 - 11 - 12 2366 60

View Full Product Details