Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024952.2 | NP_001020123.1 | 17 - 18 | 3388 | 71 |
GenBank mRNA | AK139648.1 | - | 17 - 18 | 3360 | 71 |
AK173335.1 | - | 17 - 18 | 3334 | 71 | |
AY948287.1 | - | 17 - 18 | 3329 | 71 | |
BC138663.1 | - | 16 - 17 | 3112 | 71 |