Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134741.1 | NP_001128213.1 | 7 - 8 | 1158 | 97 |
NM_001277730.1 | NP_001264659.1 | 7 - 8 | 864 | 97 | |
GenBank mRNA | AK132674.1 | - | 7 - 8 | 1144 | 97 |
AK133026.1 | - | 5 - 6 | 529 | 97 | |
BC099972.1 | - | 7 - 8 | 864 | 97 |