Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026037.3 | NP_080313.2 | 4 - 5 | 497 | 114 |
GenBank mRNA | AB297383.1 | - | 4 - 5 | 396 | 114 |
AB305046.1 | - | 4 - 5 | 396 | 114 | |
AU067514.1 | - | 4 - 5 | 497 | 114 |