Assay Design



Gene Transcripts

Gene Symbol
Ppp1r12b
Entrez Gene ID
329251
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 12B
Gene Aliases
1810037O03Rik, 9530009M10Rik, AI132431, Mypt2
Chromosome Location
Chr.1: 134754665 - 134955965 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081307.1 NP_001074776.1 22 - 23 2954 96
GenBank mRNA AK039445.1 - 10 - 11 1015 96
BC145746.1 - 22 - 23 2865 96

View Full Product Details