Assay Design



Gene Transcripts

Gene Symbol
Acot12
Entrez Gene ID
74156
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1
Chromosome Location
Chr.13: 91741511 - 91786148 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028790.3 NP_083066.1 8 - 9 936 67
GenBank mRNA AB078618.1 - 8 - 9 933 67
AK004905.1 - 8 - 9 937 67
AK033663.1 - 8 - 9 936 67
AK034622.1 - 8 - 9 940 67
BC025852.1 - 8 - 9 889 67
BC132050.1 - 8 - 9 967 67
BC137996.1 - 8 - 9 967 67

View Full Product Details