Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008006.2 | NP_032032.1 | 2 - 3 | 475 | 119 |
GenBank mRNA | AF065903.1 | - | 2 - 3 | 278 | 119 |
AF065904.1 | - | 2 - 3 | 278 | 119 | |
AF065905.1 | - | 2 - 3 | 278 | 119 | |
AY027548.1 | - | 2 - 3 | 193 | 119 | |
AY027549.1 | - | 2 - 3 | 298 | 119 | |
AY027557.1 | - | 2 - 3 | 348 | 119 | |
AY027558.1 | - | 2 - 3 | 339 | 119 | |
AY027559.1 | - | 2 - 3 | 202 | 119 | |
M30644.1 | - | 2 - 3 | 278 | 119 |