Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159725.2 | NP_001153197.2 | 4 - 5 | 604 | 72 |
NM_008998.4 | NP_033024.1 | 4 - 5 | 835 | 72 | |
GenBank mRNA | AB232611.1 | - | 3 - 4 | 441 | 72 |
AK009707.1 | - | 4 - 5 | 605 | 72 | |
AK075591.1 | - | 4 - 5 | 836 | 72 | |
BC013170.1 | - | 4 - 5 | 684 | 72 | |
BC051071.1 | - | 4 - 5 | 811 | 72 | |
X70804.1 | - | 3 - 4 | 441 | 72 |