Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026211.2 | NP_001021382.1 | 4 - 5 | 374 | 72 |
NM_001286972.1 | NP_001273901.1 | 4 - 5 | 374 | 72 | |
NM_001286973.1 | NP_001273902.1 | 3 - 4 | 322 | 72 | |
NM_026332.4 | NP_080608.3 | 4 - 5 | 374 | 72 | |
GenBank mRNA | AK007866.1 | - | 4 - 5 | 342 | 72 |
AK008267.1 | - | 4 - 5 | 318 | 72 | |
AK010204.1 | - | 4 - 5 | 324 | 72 | |
AK012051.1 | - | 4 - 5 | 326 | 72 | |
BC020327.1 | - | 4 - 5 | 307 | 72 | |
BC024335.1 | - | 4 - 5 | 282 | 72 | |
CA469052.1 | - | 4 - 5 | 411 | 72 | |
CJ123658.1 | - | 3 - 4 | 279 | 72 | |
EL962106.1 | - | 4 - 5 | 320 | 72 |