Assay Design



Gene Transcripts

Gene Symbol
Ccdc39
Entrez Gene ID
51938
Gene Name
coiled-coil domain containing 39
Gene Aliases
4921507O14Rik, AI447426, AI844750, D3Ertd789e, b2b1304Clo, b2b1735Clo, b2b2025.1Clo
Chromosome Location
Chr.3: 33810757 - 33844322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026222.2 NP_080498.1 11 - 12 1641 154
GenBank mRNA AK014839.1 - 11 - 12 1633 154
AK030087.1 - 11 - 12 1608 154
BC138917.1 - 11 - 12 1603 154
BC138920.1 - 11 - 12 1603 154

View Full Product Details