Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205391.1 | NP_001192320.1 | 12 - 13 | 1371 | 97 |
NM_001205392.1 | NP_001192321.1 | 10 - 11 | 1266 | 97 | |
NM_029846.4 | NP_084122.2 | 11 - 12 | 1323 | 97 | |
GenBank mRNA | AB087879.1 | - | 10 - 11 | 1077 | 97 |
AB087880.1 | - | 11 - 12 | 1134 | 97 | |
AB087881.1 | - | 12 - 13 | 1182 | 97 | |
AB476646.1 | - | 12 - 13 | 1182 | 97 | |
AK005181.1 | - | 12 - 13 | 1296 | 97 | |
AK030983.1 | - | 11 - 12 | 1264 | 97 | |
AK131120.1 | - | 10 - 11 | 2109 | 97 | |
AK155358.1 | - | 10 - 11 | 1202 | 97 | |
AK163246.1 | - | 10 - 11 | 1205 | 97 | |
AK170073.1 | - | 10 - 11 | 1200 | 97 | |
BC033362.1 | - | 2 - 3 | 644 | 97 | |
BC049122.1 | - | 12 - 13 | 1296 | 97 |