Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028916.4 | NP_083192.2 | 2 - 3 | 321 | 153 |
GenBank mRNA | AK016714.1 | - | 2 - 3 | 280 | 153 |
AK029501.1 | - | 2 - 3 | 287 | 153 | |
BC120659.1 | - | 2 - 3 | 256 | 153 | |
BC125625.1 | - | 2 - 3 | 256 | 153 |