Assay Design



Gene Transcripts

Gene Symbol
Efhc2
Entrez Gene ID
74405
Gene Name
EF-hand domain (C-terminal) containing 2
Gene Aliases
4933407D04Rik, mRib72-2
Chromosome Location
Chr.X: 17132049 - 17320461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028916.4 NP_083192.2 3 - 4 463 79
GenBank mRNA AK016714.1 - 3 - 4 422 79
AK029501.1 - 3 - 4 429 79
BC120659.1 - 3 - 4 398 79
BC125625.1 - 3 - 4 398 79

View Full Product Details