Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028916.4 | NP_083192.2 | 4 - 5 | 687 | 64 |
GenBank mRNA | AK016714.1 | - | 4 - 5 | 646 | 64 |
AK029501.1 | - | 4 - 5 | 653 | 64 | |
BC120659.1 | - | 4 - 5 | 622 | 64 | |
BC125625.1 | - | 4 - 5 | 622 | 64 |