Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028916.4 | NP_083192.2 | 6 - 7 | 1052 | 74 |
GenBank mRNA | AK016714.1 | - | 6 - 7 | 74 | |
AK029501.1 | - | 6 - 7 | 1018 | 74 | |
BC120659.1 | - | 6 - 7 | 987 | 74 | |
BC125625.1 | - | 6 - 7 | 987 | 74 |