Assay Design



Gene Transcripts

Gene Symbol
Usp9x
Entrez Gene ID
22284
Gene Name
ubiquitin specific peptidase 9, X chromosome
Gene Aliases
5730589N07Rik, AA407302, AA407699, AL022658, AL022749, Dffrx, FAF-X, Fafl
Chromosome Location
Chr.X: 13070997 - 13173328 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009481.2 NP_033507.2 7 - 8 1168 152
GenBank mRNA AK028443.1 - 7 - 8 1153 152
AK049283.1 - 8 - 9 1281 152
DQ086491.1 - 7 - 8 1129 152
U67874.1 - 7 - 8 1129 152

View Full Product Details