Assay Design
Gene Transcripts
- Gene Symbol
- Btnl9
- Entrez Gene ID
-
237754
- Gene Name
- butyrophilin-like 9
- Gene Aliases
-
B430208I01, Btn3, D330012D11Rik
- Chromosome Location
-
Chr.11: 49168325 - 49187089 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY177687.1 |
- |
9 - 10 |
1349 |
73 |
View Full Product Details