Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136070.1 | NP_001129542.1 | 12 - 13 | 1513 | 60 |
NM_010709.3 | NP_034839.2 | 12 - 13 | 1513 | 60 | |
GenBank mRNA | AK030312.1 | - | 12 - 13 | 1524 | 60 |
AK034115.1 | - | 12 - 13 | 1485 | 60 | |
AK036154.1 | - | 12 - 13 | 1509 | 60 | |
AK046539.1 | - | 12 - 13 | 1525 | 60 | |
AK047005.1 | - | 11 - 12 | 1358 | 60 | |
AK082748.1 | - | 12 - 13 | 1528 | 60 | |
AK083638.1 | - | 12 - 13 | 1473 | 60 | |
AK139052.1 | - | 12 - 13 | 1513 | 60 | |
AK161269.1 | - | 12 - 13 | 1496 | 60 | |
AK166085.1 | - | 12 - 13 | 1526 | 60 | |
AK167980.1 | - | 12 - 13 | 1509 | 60 | |
AK168120.1 | - | 12 - 13 | 1479 | 60 | |
AK170252.1 | - | 12 - 13 | 1540 | 60 | |
AK170448.1 | - | 12 - 13 | 1525 | 60 | |
AK171115.1 | - | 12 - 13 | 1491 | 60 | |
BC023709.1 | - | 12 - 13 | 1466 | 60 | |
BC024501.1 | - | 7 - 8 | 827 | 60 | |
BC025036.1 | - | 12 - 13 | 1469 | 60 | |
U58337.1 | - | 12 - 13 | 1398 | 60 |