Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163579.1 | NP_001157051.1 | 16 - 17 | 2828 | 165 |
NM_001163580.1 | NP_001157052.1 | 16 - 17 | 2780 | 165 | |
NM_028915.3 | NP_083191.2 | 16 - 17 | 2828 | 165 | |
GenBank mRNA | AK004646.1 | - | 16 - 17 | 2829 | 165 |
AK029563.1 | - | 16 - 17 | 2746 | 165 | |
AK030800.1 | - | 8 - 9 | 1422 | 165 | |
AK078433.1 | - | 5 - 6 | 825 | 165 | |
AK173256.1 | - | 16 - 17 | 2756 | 165 | |
BC061202.1 | - | 16 - 17 | 2736 | 165 | |
BC138734.1 | - | 16 - 17 | 2802 | 165 | |
BC138735.1 | - | 16 - 17 | 2802 | 165 |