Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163579.1 | NP_001157051.1 | 1 - 2 | 234 | 117 |
NM_001163580.1 | NP_001157052.1 | 1 - 2 | 234 | 117 | |
NM_028915.3 | NP_083191.2 | 1 - 2 | 234 | 117 | |
GenBank mRNA | AK004646.1 | - | 1 - 2 | 235 | 117 |
AK029563.1 | - | 1 - 2 | 151 | 117 | |
AK173256.1 | - | 1 - 2 | 162 | 117 | |
BC061202.1 | - | 1 - 2 | 190 | 117 | |
BC099680.1 | - | 1 - 2 | 198 | 117 | |
BC138734.1 | - | 1 - 2 | 208 | 117 | |
BC138735.1 | - | 1 - 2 | 208 | 117 |