Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163579.1 | NP_001157051.1 | 4 - 5 | 679 | 120 |
NM_001163580.1 | NP_001157052.1 | 4 - 5 | 679 | 120 | |
NM_028915.3 | NP_083191.2 | 4 - 5 | 679 | 120 | |
GenBank mRNA | AK004646.1 | - | 4 - 5 | 680 | 120 |
AK016559.1 | - | 4 - 5 | 618 | 120 | |
AK029563.1 | - | 4 - 5 | 596 | 120 | |
AK173256.1 | - | 4 - 5 | 607 | 120 | |
BC061202.1 | - | 4 - 5 | 635 | 120 | |
BC099680.1 | - | 4 - 5 | 643 | 120 | |
BC138734.1 | - | 4 - 5 | 653 | 120 | |
BC138735.1 | - | 4 - 5 | 653 | 120 |