Assay Design



Gene Transcripts

Gene Symbol
Mapkapk2
Entrez Gene ID
17164
Gene Name
MAP kinase-activated protein kinase 2
Gene Aliases
AA960234, MAPKAP-K2, MK-2, MK2, Rps6kc1
Chromosome Location
Chr.1: 131053700 - 131097543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008551.1 NP_032577.1 7 - 8 866 76
GenBank mRNA BC024559.1 - 3 - 4 235 76
BC063064.1 - 7 - 8 866 76
X76850.1 - 7 - 8 845 76

View Full Product Details