Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110150.1 | NP_001103620.1 | 1 - 2 | 101 | 74 |
GenBank mRNA | AK004760.1 | - | 1 - 2 | 101 | 74 |
AK087959.1 | - | 1 - 2 | 65 | 74 | |
AK155322.1 | - | 1 - 2 | 142 | 74 | |
BC006629.1 | - | 1 - 2 | 50 | 74 |