Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178628.5 | NP_848743.1 | 5 - 6 | 913 | 89 |
GenBank mRNA | AK046919.1 | - | 4 - 5 | 643 | 89 |
AK050339.1 | - | 5 - 6 | 750 | 89 | |
AK140185.1 | - | 4 - 5 | 657 | 89 | |
BC050784.1 | - | 4 - 5 | 642 | 89 |