Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008795.2 | NP_032821.1 | 8 - 9 | 952 | 61 |
GenBank mRNA | AK004998.1 | - | 8 - 9 | 896 | 61 |
AK171569.1 | - | 8 - 9 | 733 | 61 | |
BC119617.1 | - | 9 - 10 | 828 | 61 | |
BC119618.1 | - | 3 - 4 | 219 | 61 | |
X69026.1 | - | 8 - 9 | 969 | 61 |