Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007923.2 | NP_031949.2 | 4 - 5 | 1535 | 62 |
GenBank mRNA | AK041964.1 | - | 2 - 3 | 217 | 62 |
AK156537.1 | - | 4 - 5 | 1532 | 62 | |
BC004798.1 | - | 2 - 3 | 780 | 62 | |
BC041679.1 | - | 2 - 3 | 768 | 62 | |
BC144844.1 | - | 4 - 5 | 1285 | 62 | |
BC145795.1 | - | 4 - 5 | 1285 | 62 |