Assay Design
Gene Transcripts
- Gene Symbol
- Cetn4
- Entrez Gene ID
-
207175
- Gene Name
- centrin 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 37308627 - 37312446 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK166515.1 |
- |
2 - 3 |
|
82 |
View Full Product Details