Assay Design
Gene Transcripts
- Gene Symbol
- Kcnmb2
- Entrez Gene ID
-
72413
- Gene Name
- potassium large conductance calcium-activated channel, subfamily M, beta member 2
- Gene Aliases
-
2700049B16Rik, 3110031N04Rik
- Chromosome Location
-
Chr.3: 31902132 - 32200180 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012400.1 |
- |
1 - 2 |
157 |
73 |
View Full Product Details