Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025316.2 | NP_079592.2 | 2 - 3 | 226 | 85 |
GenBank mRNA | AK002279.1 | - | 2 - 3 | 228 | 85 |
AK004372.1 | - | 2 - 3 | 226 | 85 | |
AK013223.1 | - | 2 - 3 | 228 | 85 | |
AK018562.1 | - | 2 - 3 | 233 | 85 | |
AK088061.1 | - | 2 - 3 | 235 | 85 | |
BC025155.1 | - | 2 - 3 | 219 | 85 |