Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144987.1 | NP_001138459.1 | 8 - 9 | 2074 | 69 |
NM_001144988.1 | NP_001138460.1 | 7 - 8 | 1687 | 69 | |
NM_144895.2 | NP_659144.1 | 8 - 9 | 1858 | 69 | |
GenBank mRNA | AK031545.1 | - | 7 - 8 | 1413 | 69 |
AK035081.1 | - | 6 - 7 | 1546 | 69 | |
AK036569.1 | - | 6 - 7 | 1010 | 69 | |
AK087388.1 | - | 8 - 9 | 1827 | 69 | |
AK129171.1 | - | 7 - 8 | 1666 | 69 | |
AK147760.1 | - | 8 - 9 | 2075 | 69 | |
AK159951.1 | - | 8 - 9 | 1859 | 69 | |
AK193708.1 | - | 1 - 2 | 37 | 69 | |
AK201628.1 | - | 1 - 2 | 37 | 69 | |
BC022921.1 | - | 8 - 9 | 1826 | 69 |