Assay Design



Gene Transcripts

Gene Symbol
Arhgef26
Entrez Gene ID
622434
Gene Name
Rho guanine nucleotide exchange factor (GEF) 26
Gene Aliases
4631416L12Rik, 8430436L14Rik, AI303572, AW491953, SGEF
Chromosome Location
Chr.3: 62338777 - 62462221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081295.1 NP_001074764.1 7 - 8 2481 76
GenBank mRNA AK144976.1 - 7 - 8 1987 76

View Full Product Details