Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005509.2 | NP_001005509.1 | 10 - 11 | 1614 | 98 |
GenBank mRNA | AK050971.1 | - | 10 - 11 | 1331 | 98 |
AK078519.1 | - | 10 - 11 | 1393 | 98 | |
AK145275.1 | - | 10 - 11 | 1570 | 98 | |
BC030447.1 | - | 9 - 10 | 1283 | 98 | |
BC082557.1 | - | 10 - 11 | 1419 | 98 |