Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005509.2 | NP_001005509.1 | 9 - 10 | 1049 | 147 |
GenBank mRNA | AK044865.1 | - | 3 - 4 | 1096 | 147 |
AK050971.1 | - | 9 - 10 | 766 | 147 | |
AK078519.1 | - | 9 - 10 | 828 | 147 | |
AK145275.1 | - | 9 - 10 | 1005 | 147 | |
BC030447.1 | - | 8 - 9 | 718 | 147 | |
BC082557.1 | - | 9 - 10 | 854 | 147 |