Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027534.2 | NP_081810.1 | 9 - 10 | 971 | 108 |
GenBank mRNA | AK018155.1 | - | 10 - 11 | 946 | 108 |
AK170064.1 | - | 9 - 10 | 854 | 108 | |
AY634684.1 | - | 9 - 10 | 880 | 108 | |
BC023820.1 | - | 9 - 10 | 961 | 108 |