Assay Design
Gene Transcripts
- Gene Symbol
- Scly
- Entrez Gene ID
-
50880
- Gene Name
- selenocysteine lyase
- Gene Aliases
-
9830169H08, A930015N15Rik, SCL, Scly1, Scly2
- Chromosome Location
-
Chr.1: 91298295 - 91321080 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK020864.1 |
- |
1 - 2 |
62 |
84 |
View Full Product Details